Recombinant Protein;25TESTS;PE-Labeled Human Her2, His Tag (HE2-HP2E3) is produced via conjugation of PE to Human Her2, His Tag under optimal conditions. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. ;5e5 of anti-Her2 CAR-293 cells were stained with 100 L of 1:25 dilution (4 L stock solution in 100 L FACS buffer) of PE-Labeled Human Her2, His Tag (Cat. No. HE2-HP2E3) and negative control protein respectively. PE signal was used to evaluate the binding activity (QC tested).