Learn More
SignalChem Biotech AMPK (A1/B1/G2), Active 50ug
Numéro de catalogue. 502573611
Encompass
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.
Numéro de catalogue. 502573611 Fournisseur SignalChem Biotech Code fournisseur P5510G50
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.