missing translation for 'onlineSavingsMsg'
Learn More

PD 153035 Hydrochloride 97.0+%, TCI America™

Catalog No. B494525MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B494525MG 25 mg
B4945100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B494525MG Supplier TCI America Supplier No. B494525MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 183322-45-4
Molecular Formula C16H15BrClN3O2
Molecular Weight (g/mol) 396.669
MDL Number MFCD01074805
InChI Key ZJOKWAWPAPMNIM-UHFFFAOYSA-N
Synonym pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride
PubChem CID 11246488
IUPAC Name N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride
SMILES COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl

Specifications

Melting Point 265°C
Quantity 25 mg
Formula Weight 396.67
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material PD 153035 Hydrochloride
TSCA No
Recommended Storage Freezer (-20 °C)