missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PCR™ Cooler

Catalog No. 01670465
Change view
Click to view available options
Color:
Assorted
Blue
Pink, Purple
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01670465 Pink, Purple
01670466 Blue
01670459 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01670465 Supplier Fisherbrand™ Supplier No. 120727F2
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ice-Free Cooling System for PCR Preparation and Storage.

  • The PCR Cooler is ideal for thermosensitive applications, PCR, or cell-based assays, as well as gentle sample thawing and stopping of reactions
  • The unit also provides a dependable cooling station for pipetting and sample storage
  • Modern design cools faster in freezers and stays cold longer on the bench top
  • Maintains the entire 96-Well plate at a temperature of 0°C for over an hour
  • Maintains sample temperature of less than 5°C for up to three hours (with lid off)
  • When the temperature ceiling of 6°C has been exceeded, users are signaled by a contrasting color change of the cooler or a single well
  • Alphanumeric well identification for quick location and retrieval of tubes
  • Lid doubles as a stand enables the rack to stay cold longer as there is no conductive heat transfer from surface beneath
  • Vented lid enables faster cooling samples while in freezer
  • Ergonomic textured finger grips provide a secure hold, reducing the chance of dropping the PCR rack, and makes handling and transportation easy, especially when using gloves

Specifications

Autoclavable Not Autoclavable
Color Pink, Purple
Holds Microtubes (0.2 mL)
Material Polypropylene, Polycarbonate
Positions 1
Shape Rectangular, Rounded Corners
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) 0.2 mL, 0.5 mL and assay plates
Length (Metric) 11 cm
Length (English) 4.3 in.
Width (English) 5.9 in.
Height (English) 1.9 in.
Includes 2 PCR Coolers
Product Type Microtube Rack
Shelves None
Height (Metric) 4.8 cm
Array 12 x 8
Width (Metric) 14.8 cm
Show More Show Less