missing translation for 'onlineSavingsMsg'
Learn More

PBS, 10X Solution, Molecular Biology Grade

For biological research. Affymetrix/USB™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.

$88.27 - $343.89

Specifications

pH 7.4
Physical Form Solution
Formulation 80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ75889AE
View Documents
Thermo Fisher Scientific
J75889AE
100 mL
Each for $88.27
Only null left
 
AAJ75889K2
View Documents
Thermo Fisher Scientific
J75889K2
1 L
Each for $191.45
Only null left
 
AAJ75889K8
View Documents
Thermo Fisher Scientific
J75889K8
5 L
Each for $343.89
Only null left
 
Description

Description

  • Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
  • Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
  • Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
Specifications

Specifications

7.4
Solution
80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only