Learn More
PBS, 10X Solution, Molecular Biology Grade
For biological research. Affymetrix/USB™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.
Supplier:  Thermo Fisher Scientific J75889K8
 View more versions of this product
Catalog No. AAJ75889K8
Only null left
Explore available promotions
Description
Description
  • Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
  • Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
  • Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
Specifications
Specifications
7.4
Solution
5 L
80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O
Product Suggestions
Product Suggestions