Learn More
Restek™ Parker Balston Maintenance Kit for UHPN2 Series Nitrogen Generators
Catalog No. 06711710
Encompass
Change view
Click to view available options
For Use With (Equipment):
UHPN2-1100, 76-94
Catalog No. For Use With (Equipment)
06711710 UHPN2-1100, 76-94
1 options
Catalog No. 06711710 Supplier Restek™ Supplier No. RK 21655
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Safe, reliable, and requires low maintenance
  • Turns compressed air into ultra-pure nitrogen (up to 99.5%)
  • Maintenance kits include replacement filters
  • Models N2-04, N2-14, N2-22, and N2-35 require no electricity
Specifications
Certifications/Compliance CE
For Use With (Equipment) UHPN2-1100, 76-94
Includes Prefilters and final filter
For Use With (Application) Turns compressed air into ultra-pure nitrogen
Type Maintenance Kit for Models UHPN2-1100, 76-94