missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline Hydrochloride 95.0+%, TCI America™

Catalog No. P0599500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0599500G 500 g
P059925G 25 g
P0599100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0599500G Supplier TCI America Supplier No. P0599500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 270°C
Color Black-Green
Quantity 500 g
Formula Weight 323.82
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline Hydrochloride
RTECSNumber CX9850100
TSCA Yes