missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraformaldehyde MP Biomedicals
Specifications
Specifications
| Density | 1.4 g/mL |
| Quantity | 1 kg |
| Formula Weight | 90.1 |
| Physical Form | Powder |
| Chemical Name or Material | Paraformaldehyde |