missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS

Catalog No. AAJ61899AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61899AK 250 mL
AAJ61899AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61899AK Supplier Thermo Scientific Chemicals Supplier No. J61899AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Paraformaldehyde, CH2O, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane, 500mL, CHEBI:16842, H317-H318-H351 | CH2O | 30.026 g/mol

Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
Chemical Name or Material Paraformaldehyde
Hazard Category Serious eye damage/eye irritation (category 1), Carcinogenicity (category 2), Skin sensitisation (category 1)
Hazard Statement Causes serious eye damage, Suspected of causing cancer, May cause an allergic skin reaction
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only