Learn More
GA International Paper Labels for Thermal-Transfer Printers 0.354"" / 9mm Circle Green Seafoam From -20C to +120C (-4F to +248F)
Numéro de catalogue. 5000572072
Encompass
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Numéro de catalogue. 5000572072 Fournisseur GA International Code fournisseur GPB61NPNOTC12GS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.