missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papanicolaou Stain, OG-6 Formula, Modified, Ricca Chemical
Counterstain for use with Hematoxylin Stains in Papanicolaou Staining
Specifications
| Chemical Name or Material | Papanicolaou Stain |
|---|---|
| Name Note | OG-6 Formula |
| CAS | 7732-18-5 , 1969-15-8 , 12067-99-1 , 7732-18-5 |
| CAS Min % | 80.0, 17.64, 0.35, 0.02 |
| CAS Max % | 83.0, 18.36, 0.37, 0.02 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
55101
|
N/A | 4 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
551032
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
551016
|
N/A | 500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| Papanicolaou Stain | |
| 7732-18-5 , 1969-15-8 , 12067-99-1 , 7732-18-5 | |
| 83.0, 18.36, 0.37, 0.02 |
| OG-6 Formula | |
| 80.0, 17.64, 0.35, 0.02 |