missing translation for 'onlineSavingsMsg'
Learn More

Papanicolaou Stain, OG-6 Formula, Modified, Ricca Chemical

Catalog No. 551016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
551016 500 mL Natural Poly Bottle
55101 4 L Natural Poly Bottle
551032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 551016 Supplier Ricca Chemical Company Supplier No. 551016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Counterstain for use with Hematoxylin Stains in Papanicolaou Staining

Specifications

Chemical Name or Material Papanicolaou Stain
Name Note OG-6 Formula
CAS 22059-21-8, 7732-18-5, 1969-15-8, 12067-99-1, 7732-18-5
CAS Min % 80.0, 17.64, 0.35, 0.02
CAS Max % 83.0, 18.36, 0.37, 0.02
Packaging Natural Poly Bottle
Color Orange
Quantity 500 mL