Learn More
Papanicolaou Stain, OG-6 Formula, Modified, Ricca Chemical
Counterstain for use with Hematoxylin Stains in Papanicolaou Staining
Supplier:  Ricca Chemical Company 551032
 View more versions of this product
Catalog No. 551032
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Chemical Identifiers
80.0, 17.64, 0.35, 0.02
83.0, 18.36, 0.37, 0.02
7732-18-5, 1969-15-8, 12067-99-1, 7732-18-5
Papanicolaou Stain, OG-6 Formula
Specifications
Papanicolaou Stain
80.0, 17.64, 0.35, 0.02
7732-18-5 , 1969-15-8 , 12067-99-1 , 7732-18-5
Orange
OG-6 Formula
83.0, 18.36, 0.37, 0.02
Natural Poly Bottle
1 L
Product Suggestions
Product Suggestions