missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Papain, for Biochemistry

Catalog No. AC416765000
Change view
Click to view available options
Quantity:
10g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416765000 500g
AC416760100 10g
AC416761000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416765000 Supplier Thermo Scientific Chemicals Supplier No. 416765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Name Papain
pH Range 4.8 to 6.2
Quantity 500g
Source Plant
Packaging Glass bottle
Form Powder
What does the unit TU/mg mean for enzyme activity versus the commonly used unit U/mg?

Papain activity is measured in terms of units of tyrosine release per mg (TU/mg), and this unit can be used interchangeably with the more general units per mg (U/mg).

RUO – Research Use Only