missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole sodium salt hydrate

Catalog No. AC460870050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460870050 5 g
AC460870010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460870050 Supplier Thermo Scientific Chemicals Supplier No. 460870050

missing translation for 'validMainframeMsg'

CAS: 718635-09-7 | C16H14F2N3NaO4S | 405.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 718635-09-7
Molecular Formula C16H14F2N3NaO4S
Molecular Weight (g/mol) 405.35
MDL Number MFCD08704580
InChI Key YNWDKZIIWCEDEE-UHFFFAOYNA-N
Synonym pantoprazole sodium, pantoprazole sodium hydrate, pantozol hydrate, protonix hydrate, c16h14f2n3nao4s.h2o, sodium pantoprazole 1-hydrate, pantoprazole sodium hydrate hplc, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
PubChem CID 23684923
IUPAC Name sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
SMILES [Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1

Specifications

Melting Point 151.0°C to 155.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information (1% in water) Clear solution
Formula Weight 405.35
Chemical Name or Material Pantoprazole sodium salt hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only