Learn More
Motic™ Panthera C2 Compound LED Microscopes
GREENER_CHOICE
The Panthera C2 is prepared for Phase Contrast with slider or turret condenser, Polarization Contrast, and Darkfield. It is even possible to integrate a ready-to-go LED fluorescence module, allowing for advanced methods without problems associated with mercury bulb usage.
$6,571.27 - $7,468.91
Specifications
Magnification Power 10X
Objective Types CCIS™ Plan UC Objectives (4X, 10X, 40X(S), 100X(S-oil))
Nosepiece Reversed Quadruple
Eyepiece WF 10X/20 mm
View More Specs
Products 2
Catalog Number Mfr. No. Head Style Lighting Type Price Quantity & Availability  
Catalog Number Mfr. No. Head Style Lighting Type Price Quantity & Availability  
01258022
GREENER_CHOICE
View Documents Promotion Details
Motic™
1100104601136
Binocular LED
Each for $6,571.27
Only null left
01258042
View Documents Promotion Details
Motic™
1100104601147
Trinocular Kohler Illumination
Each for $7,468.91
Only null left
Description
Description
  • Swiveling, 25° inclined Siedentopf head
  • Nosepiece with light tracer (Intelli Light Management)
  • LED light indicator
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built-in, low position coaxial rackless stage with right hand control
  • Focusable Abbe condenser with 0.9 numerical aperature
  • Iris diaphragm and slot
  • Diopter Adjustment: ±5
Specifications
Specifications
10X
CCIS™ Plan UC Objectives (4X, 10X, 40X(S), 100X(S-oil))
Reversed Quadruple
WF 10X/20 mm