Learn More
Motic™ Panthera C2 Compound LED Microscopes
GREENER_CHOICE
Catalog No. 01258022
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Lighting Type:
Kohler Illumination
LED
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Lighting Type
01258022 Binocular LED
01258042 Trinocular Kohler Illumination
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01258022 Supplier Motic™ Supplier No. 1100104601136
Only null left
The Panthera C2 is prepared for Phase Contrast with slider or turret condenser, Polarization Contrast, and Darkfield. It is even possible to integrate a ready-to-go LED fluorescence module, allowing for advanced methods without problems associated with mercury bulb usage.
  • Swiveling, 25° inclined Siedentopf head
  • Nosepiece with light tracer (Intelli Light Management)
  • LED light indicator
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built-in, low position coaxial rackless stage with right hand control
  • Focusable Abbe condenser with 0.9 numerical aperature
  • Iris diaphragm and slot
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types CCIS™ Plan UC Objectives (4X, 10X, 40X(S), 100X(S-oil))
Nosepiece Reversed Quadruple
Head Style Binocular
Lighting Type LED
Eyepiece WF 10X/20 mm