Learn More
Motic™ Panthera C2 Compound LED Microscopes
Description
- Swiveling, 25° inclined Siedentopf head
- Nosepiece with light tracer (Intelli Light Management)
- LED light indicator
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built-in, low position coaxial rackless stage with right hand control
- Focusable Abbe condenser with 0.9 numerical aperature
- Iris diaphragm and slot
- Diopter Adjustment: ±5
Specifications
Specifications
| Magnification Power | 10X |
| Objective Types | CCIS™ Plan UC Objectives (4X, 10X, 40X(S), 100X(S-oil)) |
| Nosepiece | Reversed Quadruple |
| Head Style | Trinocular |
| Lighting Type | Kohler Illumination |
| Eyepiece | WF 10X/20 mm |