missing translation for 'onlineSavingsMsg'
Learn More

Pancreatin U.S.P., MP Biomedicals™

Catalog No. ICN10255780
Change view
Click to view available options
Quantity:
100 g
1 kg
Catalog No. Quantity
ICN10255780 100 g
ICN10255791 1 kg
2 options

Catalog No. ICN10255780

Supplier: MP Biomedicals Inc 0210255780

Only null left

Chemical Identifiers

CAS 8049-47-6
Synonym Diastase vera

Specifications

Color Tan to White
Quantity 100 g
Physical Form Powder
Chemical Name or Material Pancreatin