missing translation for 'onlineSavingsMsg'
Learn More

Pancreatin U.S.P., MP Biomedicals™

Catalog No. ICN10255791
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10255791 1 kg
ICN10255780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10255791 Supplier MP Biomedicals Inc Supplier No. 0210255791
Only null left

Chemical Identifiers

CAS 8049-47-6
Synonym Diastase vera

Specifications

Color Tan to White
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Pancreatin