missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAN Indicator, 0.3% in Ethanol, Certified, LabChem™
Spécifications
Spécifications
| Chemical Name or Material | PAN Indicator |
| Name Note | 0.3% in Ethanol |
| CAS Max % | 89.7, 5, 5, 0.3 |
| CAS | 67-56-1, 67-63-0, 85-85-8 |
| Linear Formula | C15H11N3O |
| UN Number | UN1170 |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Formula Weight | 249.27 |
| Grade | Certified |
| Afficher plus de résultats |