missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palmitoleic acid, 99%

Catalog No. AC376912500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC376912500 250 mg Glass bottle
AC376910010 1 g Glass bottle
AC376910050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC376912500 Supplier Thermo Scientific Chemicals Supplier No. 376912500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 373-49-9 | C16H30O2 | 254.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 373-49-9
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00004437
InChI Key SECPZKHBENQXJG-FPLPWBNLSA-N
Synonym palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
PubChem CID 445638
ChEBI CHEBI:28716
IUPAC Name (Z)-hexadec-9-enoic acid
SMILES CCCCCC\C=C/CCCCCCCC(O)=O

Specifications

Melting Point 0.5°C
Density 0.8940g/mL
Boiling Point 162.0°C (0.6 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590
Quantity 250 mg
Specific Gravity 0.894
Solubility Information Solubility in water: insoluble
Formula Weight 254.41
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Palmitoleic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 206-765-9

RUO – Research Use Only