missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid 99.5+%, TCI America™

Catalog No. P11455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11455G Supplier TCI America Supplier No. P11455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-10-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00002747
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
Synonym palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
PubChem CID 985
ChEBI CHEBI:15756
IUPAC Name hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 64°C
Color White
Boiling Point 352°C
Quantity 5 g
Formula Weight 256.43
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Palmitic Acid
EINECSNumber (2)-0608
RTECSNumber RT4550000
TSCA Yes