missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, FCC, Spectrum™ Chemical

Catalog No. 18604952
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604952 50 kg Poly Bottle
18604950 500 g Poly Bottle
18604951 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604952 Supplier Spectrum Chemical Mfg Cor Supplier No. P151350KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1513, 57-10-3, C16H32O2

Spectrum™ Chemical Palmitic Acid, FCC adds texture and "mouth feel" to processed foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-10-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00002747
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
IUPAC Name hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(O)=O

Specifications

CAS Max % 1
Boiling Point 350°C
Ignition Residue 0.001
Melting Range 61°C to 64°C
Packaging Poly Bottle
Quantity 50 kg
Saponification Value 205-221
Grade FCC
Physical Form Crystals or Flakes
Chemical Name or Material Palmitic Acid