missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitic Acid, >99%, MP Biomedicals™
Chemical Identifiers
| CAS | 57-10-3 |
|---|---|
| Molecular Formula | C16H32O2 |
| Molecular Weight (g/mol) | 256.43 |
| MDL Number | MFCD00002747 |
| InChI Key | IPCSVZSSVZVIGE-UHFFFAOYSA-N |
| Synonym | palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid |
| PubChem CID | 985 |
| ChEBI | CHEBI:15756 |
| IUPAC Name | hexadecanoic acid |
| SMILES | CCCCCCCCCCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10090510
|
N/A | 10 g |
N/A
|
|
|||||
|
ICN10090583
|
N/A | 250 g |
N/A
|
|
|||||
Chemical Identifiers
| C16H32O2 | |
| MFCD00002747 | |
| palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid | |
| CHEBI:15756 | |
| CCCCCCCCCCCCCCCC(O)=O |
Specifications
| 57-10-3 | |
| 1.4273g/mL | |
| C16H32O2 | |
| palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid | |
| CCCCCCCCCCCCCCCC(O)=O | |
| 256.43 | |
| CHEBI:15756 | |
| Crystalline Powder |
| White | |
| >99% | |
| MFCD00002747 | |
| IPCSVZSSVZVIGE-UHFFFAOYSA-N | |
| hexadecanoic acid | |
| 985 | |
| 256.4 | |
| Palmitic acid |