missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, >99%, MP Biomedicals™

Catalog No. ICN10090510
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10090510 10 g
ICN10090583 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10090510 Supplier MP Biomedicals Inc Supplier No. 0210090510

Chemical Identifiers

CAS 57-10-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00002747
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
Synonym palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
PubChem CID 985
ChEBI CHEBI:15756
IUPAC Name hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(O)=O

Specifications

Color White
Density 1.4273g/mL
Assay Percent Range >99%
Quantity 10 g
Formula Weight 256.4
Physical Form Crystalline Powder
Chemical Name or Material Palmitic acid