missing translation for 'onlineSavingsMsg'
Learn More

Palmitanilide 98,0+%, TCI America™

Numéro de catalogue. P084425G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P084425G 25 g
P0844500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P084425G Fournisseur TCI America Code fournisseur P084425G

Identifiants chimiques

CAS 6832-98-0
Molecular Formula C22H37NO
Molecular Weight (g/mol) 331.544
MDL Number MFCD00059287
InChI Key VENJCACPSJGPCM-UHFFFAOYSA-N
Synonym N-Palmitoylaniline, N-Phenylpalmitamide
PubChem CID 96171
IUPAC Name N-phénylhexadécanamide
SMILES CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1

Spécifications

Melting Point 90°C
Color Blanc
Boiling Point 284°C
Quantity 25 g
Formula Weight 331.54
Percent Purity ≥98.0% (N)
Physical Form Poudre cristalline
Chemical Name or Material Palmitanilide
EINECSNumber (3)-2698
TSCA No