missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, 97%, Thermo Scientific™

Catalog No. AC467152500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467152500 250 mg
AC467150010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467152500 Supplier Thermo Scientific Chemicals Supplier No. 467152500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10605-02-4
Molecular Formula C21 H22 Cl N O4

Specifications

Quantity 250 mg
Formula Weight 387.86
Physical Form Crystalline Powder
Color Light-yellow to Yellow to Orange
Purity ≥97.5%
Chemical Name or Material Palmatine chloride
Hazard Category Exclamation mark
Recommended Storage Normal conditions