missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Palladium On Carbon

Numéro de catalogue. 205699250G
Encompass
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
205699250G 250 g
20569910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 205699250G Fournisseur Sigma Aldrich Code fournisseur 205699250G
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Palladium on carbon (Pd/C) (10 wt. % loading, matrix activated carbon support) can be used as a catalyst in the well known reactions like hydrogenation of alkenes, alkynes, ketones, nitriles, imines, azides, nitro groups, benzenoid and heterocyclic aromatics; hydrogenolysis of cyclopropanes, benzyl derivatives, epoxides, hydrazines, and halides; dehydrogenate aromatics and deformylate aldehydes.It can also be used as:A catalyst for hydrodechlorination of dichlorodifluoromethane (CCl2F2), which is a known greenhouse gas,A catalyst for Heck reactions of aryl halides in ionic liquidsAn anode catalyst for the fabrication of direct formic acid fuel cells (DFAFC)A cost-effective electrocatalyst for the oxidation of methanolIn the catalytic hydrogenation of chemical hydrides for the production of hydrogenA catalyst in Suzuki-Mayaura reaction

Spécifications

Molecular Formula Pd
Linear Formula Pd
Quantity 250 g
MDL Number MFCD03457879
Synonym Pd/C
Molecular Weight (g/mol) 106.42 g/mol