missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Palladium On Carbon
Description
Useful reagent for debenzylations and decarboxybenzylations.Noblyst is a registered trademark of Evonik Degussa GmbH
Spécifications
Spécifications
| Molecular Formula | Pd |
| Linear Formula | Pd |
| MDL Number | MFCD03457879 |
| Quantity | 250 g |
| Synonym | Pd/C |
| Molecular Weight (g/mol) | 106.42 g/mol |