missing translation for 'onlineSavingsMsg'
Learn More

Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™

Catalog No. AA45290AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45290AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45290AK Supplier Thermo Scientific Chemicals Supplier No. 045290AK

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD09910543

Specifications

Physical Form Liquid
Concentration 0.075% Pd & 0.05% Mg(NO3)2 in 2% HNO3
UN Number UN3264
Quantity 250 mL
Grade Specpure
Chemical Name or Material Palladium nitrate & Magnesium nitrate Matrix Modifier Solution
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only