Learn More
Palladium(II) chloride, Premion™, 99.999% (metals basis), Pd 59.5% min
Description
Palladium(II) chloride is used to remove the stain present stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tatooing of leukomas in thecornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 675°C (decomposition) |
| Color | Red to Brown |
| Physical Form | Powder |
| Quantity | 1 g |
| Merck Index | 14,6990 |
| Solubility Information | Soluble in cold water,organic solvents and hydrochloric acid. |
| IUPAC Name | dichloropalladium |
| Formula Weight | 177.31 |
| Percent Purity | 99.999% |
| Grade | Premion |
| Show More |
Safety and Handling
| Hazard Category | H290-H302-H317-H318 |
| Hazard Statement | GHS H Statement H318-H290-H317 Causes serious eye damage. May be corrosive to metals. May cause an allergic skin reaction. |
| Precautionary Statement | P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 231-596-2 |
| RTECSNumber | RT3500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only