missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide, 99%

Catalog No. AA3949603
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3949603 1 g
AA3949601 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3949603 Supplier Thermo Scientific Chemicals Supplier No. 03949603
Only null left

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol

Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
SMILES [OH-].[OH-].[Pd++]

Specifications

Color Black
Physical Form Powder
Quantity 1 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
IUPAC Name palladium(2+);dihydroxide
Formula Weight 140.42
Percent Purity 99%
Odor Odorless
Chemical Name or Material Palladium hydroxide
EINECSNumber 235-219-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only