missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium AA Standard, 1000 ppm Pd in 3% HCl, Ricca Chemical
$463.23 - $463.23
Chemical Identifiers
| CAS | 7440-05-3 |
|---|---|
| Molecular Formula | Pd |
| Molecular Weight (g/mol) | 106.42 |
| MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
| InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
| PubChem CID | 23938 |
| ChEBI | CHEBI:33363 |
| IUPAC Name | palladium |
| SMILES | [Pd] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
APD1KH-100
|
Ricca Chemical Company
APD1KH100 |
100 mL | Natural LDPE Bottle |
Each for $463.23
|
|
||||
Chemical Identifiers
| 7440-05-3 | |
| 106.42 | |
| KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
| CHEBI:33363 | |
| [Pd] |
| Pd | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| 23938 | |
| palladium |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Pd | |
| 95.0,5.63,0.1 | |
| Liquid | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| Approximately 0°C | |
| [Pd] | |
| 106.42 | |
| 23938 | |
| Palladium AA Standard |