missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel, 99.5+%

Catalog No. AAJ62734ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62734ME 500 mg
AAJ62734MC 100 mg
AAJ6273403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62734ME Supplier Thermo Scientific Chemicals Supplier No. J62734ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Paclitaxel (CAS # 33069-62-4) is a microtubule inhibitor produced from the extract of the Pacific yew tree. | CAS: 33069-62-4 | C47H51NO14 | 853.92 g/mol

Promotes assembly and inhibits disassembly of microtubules (anti-cancer agent)Paclitaxel is widely used in chemotherapeutic compound. It is used to treat certain types of cancer such as ovarian cancer, breast cancer, lung cancer and pancreatic cancer among others. It interferes with the growth of cancer cells and slows down their growth and spread in the body. It is also used to treat AIDS-related Kaposi's sarcoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Paclitaxel is a tetracyclic diterpenoid isolated from the Pacific yew, Taxus brevifolia
  • Paclitaxel acts as an antineoplastic agent by binding tubulin and preventing cell division
  • Paclitaxel may also bind Bcl-2 and lead to disinhibition of apoptosis

Application

  • Paclitaxel is used in the study of lung, breast, and ovarian cancers; it may be useful in the research of other cancer types
  • It is suitable for cell culture, biochemistry, and molecular biology applications

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14
Molecular Weight (g/mol) 853.92
MDL Number MFCD00869953
InChI Key RCINICONZNJXQF-VAZQATRQSA-N
Synonym paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
PubChem CID 133640187
IUPAC Name (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213°C (decomposition)
Color White
Quantity 500 mg
Beilstein 1420457
Merck Index 14,6982
Solubility Information Soluble in DMSO (200mg/ml) or ethanol (40mg/ml); Soluble in dimethylsulfoxide,methanol,ethanol,acetonitrile,cremophor and dimehtylsulfamide; Sparingly soluble in aqueous buffer.
Formula Weight 853.91
Percent Purity ≥99.5%
Physical Form Crystalline Solid
Chemical Name or Material Paclitaxel
Hazard Category H302+H312+H332-H318-H334-H335-H361
Hazard Statement GHS H Statement
H334-H341-H361-H318-H302-H312-H332-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P342+P311-P363-P501c
RTECSNumber DA8340700
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only