missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Wide Mouth HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000121
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000121 1 L
S000117 60 mL
S000118 125 mL
S000119 250 mL
S000120 500 mL
S000116 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S000121 Supplier Eisco Labs Supplier No. CH5466PK12

Store and handle lab liquids with confidence using Eisco Labs wide-mouth translucent HDPE bottles, each fitted with a polypropylene cap for leakproof performance, chemical resistance, and easy pouring in demanding lab or industrial settings.

  • Reagent bottles: Includes twelve translucent wide-mouth bottles made of HDPE, featuring polypropylene caps, ideal for reagent storage and lab use.
  • Excellent chemical resistance: Made with HDPE material that resists a wide variety of chemicals, ensuring reliable containment for diverse applications in laboratories and industries.
  • Temperature resistance: Functional in a range from -100°C to 120°C, these bottles are designed for safe handling of solutions in cold and moderately heated conditions.
  • Leakproof design: Rigorously tested to prevent leaks, these bottles offer secure transportation and storage for liquids and reagents.
  • Ergonomic and translucent: Wide-mouth design ensures easy filling and pouring. Translucent material allows visibility of contents for efficient handling.

Specifications

Capacity (Metric) 1 L
Color Translucent
Diameter (Metric) Inner Neck 53 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 198 mm
Diameter (Metric) 93.5 mm
Autoclavable Not Autoclavable
Capacity (English) 33.8 oz.
Product Type Reagent Bottle
Show More Show Less