Learn More
Thermo Scientific™ Pacific Hemostasis™ Factor Assays
Catalog No. 23639012
Change view
Click to view available options
Type:
Factor VIII Depleted Plasma
Catalog No. Type
23639012 Factor VIII Depleted Plasma
1 options
Catalog No. 23639012 Supplier Thermo Scientific™ Supplier No. 10 0806
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fisherbrand™ Pacific Hemostasis Factor Assays are used for the detection and quantification of factor deficiencies in patient plasmas.
Detect and quantify factor deficiencies in patient plasmas with Thermo Scientific™ Pacific Hemostasis™ Factor Assays.
Pacific Hemostasis Factor Assays include a complete line of lyophilized congenital factor deficient plasmas for performing factor assays.
  • All factor deficient plasmas are prepared from congenitally deficient patient donors or immunodepleted plasma, with the exception of Factor II.
  • Factor II Deficient Plasma is a mix of human and bovine plasma.
  • Plasmas are packaged 10 × 1mL; Diluting Fluid is packaged 2 × 100mL.
Specifications
Type Factor VIII Depleted Plasma