missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
Spécifications
Spécifications
| Antigène | P4HA2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | P4HA2 |
| Numéro d’acquisition du gène | O15460 |
| Alias du gène | 4-PH alpha 2, 4-PH alpha-2, C-P4Halpha(II), collagen prolyl 4-hydroxylase alpha(II), procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II, procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2, prolyl 4-hydr |
| Symboles de gène | P4HA2 |
| Afficher plus de résultats |