missing translation for 'onlineSavingsMsg'
Learn More

P2RY14 Mouse anti-Human, Clone:5A3F1, Abnova™

Numéro de catalogue. 89334016
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89334016 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89334016 Fournisseur Abnova Corporation Code fournisseur MAB17592
Il en reste null

Mouse monoclonal antibody raised against recombinant human P2RY14.

The product of this gene belongs to the family of G-protein coupled receptors, which contains several receptor subtypes with different pharmacological selectivity for various adenosine and uridine nucleotides. This receptor is a P2Y purinergic receptor for UDP-glucose and other UDP-sugars coupled to G-proteins. It has been implicated in extending the known immune system functions of P2Y receptors by participating in the regulation of the stem cell compartment, and it may also play a role in neuroimmune function. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]

Spécifications

Antigen P2RY14
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5A3F1
Conjugate Unconjugated
Description purinergic receptor P2Y, G-protein coupled, 14
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene P2RY14
Gene Alias GPR105, KIAA0001, P2Y14
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid of human P2RY14 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 9934
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats