missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylthiourea 98.0+%, TCI America™

Catalog No. T06565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06565G Supplier TCI America Supplier No. T06565G

Chemical Identifiers

CAS 622-52-6
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD00041190
InChI Key VXLFMCZPFIKKDZ-UHFFFAOYSA-N
Synonym p-tolylthiourea, 1-p-tolyl thiourea, 1-p-tolylthiourea, 1-4-methylphenyl thiourea, n-4-methylphenyl thiourea, 4-methylphenyl thiourea, urea, 2-thio-1-p-tolyl, n-p-tolylthiourea, p-tolylthiocarbamide, thiourea, 4-methylphenyl
PubChem CID 722833
IUPAC Name (4-methylphenyl)thiourea
SMILES CC1=CC=C(NC(N)=S)C=C1

Specifications

Melting Point 180°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 166.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolylthiourea
RTECSNumber YU3325000
TSCA No