missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™

Catalog No. AC426768000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426768000 800 mL AcroSeal™ Glass bottle
AC426761000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426768000 Supplier Thermo Scientific Chemicals Supplier No. 426768000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal, Quantity: 800mL, Packaging: AcroSeal Glass bottle, Molecular Weight: 195.34, CAS: 4294-57-9, 60-29-7, Density: 0.8660g/mL, Flash Point: -40 deg.C, Formula Weight: 195.34 | CAS: 4294-57-9 | C7H7BrMg | 195.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4294-57-9 , 60-29-7
Molecular Formula C7H7BrMg
Molecular Weight (g/mol) 195.34
MDL Number MFCD00000040
InChI Key ZRJNGFJIBZKXTP-UHFFFAOYSA-M
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide, grignard reagent
PubChem CID 3644322
IUPAC Name magnesium;methylbenzene;bromide
SMILES CC1=CC=C([Mg]Br)C=C1

Specifications

Density 0.8660g/mL
Flash Point -40°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH3C6H4MgBr
Quantity 800 mL
Specific Gravity 0.866
Formula Weight 195.34
Chemical Name or Material p-Tolylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only