missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl chlorothionoformate, 97%

Catalog No. AAB2350118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2350118 50 g
AAB2350109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2350118 Supplier Thermo Scientific Chemicals Supplier No. B2350118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 937-63-3 | C8H7ClOS | 186.65 g/mol

p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2'-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2′-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

Solubility
Miscible with common organic solvents.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 937-63-3
Molecular Formula C8H7ClOS
Molecular Weight (g/mol) 186.65
MDL Number MFCD00014466
InChI Key UNCAXIZUVRKBMN-UHFFFAOYSA-N
Synonym 4-methylphenyl chlorothioformate, o-p-tolyl chlorothioformate, p-tolyl chlorothionoformate, 4-tolyl chlorothionoformate, 4-methylphenyl chloromethanethioate, o-4-methylphenyl chlorothioformate, carbonochloridothioic acid, o-4-methylphenyl ester, o-4-methylphenyl carbonochloridothioate, chlorothioformic acid p-tolyl ester, 4-methylphenoxy methanethioyl chloride
PubChem CID 70305
IUPAC Name O-(4-methylphenyl) chloromethanethioate
SMILES CC1=CC=C(OC(Cl)=S)C=C1

Specifications

Density 1.21
Boiling Point 70°C to 72°C (1 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.574
Quantity 50 g
UN Number UN3265
Beilstein 1448986
Sensitivity Moisture sensitive
Solubility Information Miscible with common organic solvents.
Formula Weight 186.66
Percent Purity 97%
Chemical Name or Material p-Tolyl chlorothionoformate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 213-333-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only