missing translation for 'onlineSavingsMsg'
Learn More

p-Toluidine Hydrochloride 98.0+%, TCI America™

Catalog No. T0303500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0303500G 500 g
T030325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0303500G Supplier TCI America Supplier No. T0303500G

Chemical Identifiers

CAS 540-23-8
Molecular Formula C7H10ClN
Molecular Weight (g/mol) 143.61
MDL Number MFCD00035497
InChI Key JQKBUTDZZRGQDR-UHFFFAOYSA-N
PubChem CID 10891
IUPAC Name hydrogen 4-methylaniline chloride
SMILES [H+].[Cl-].CC1=CC=C(N)C=C1

Specifications

Melting Point 243°C
Quantity 500 g
UN Number 2811
Formula Weight 143.61
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluidine Hydrochloride
EINECSNumber (3)-0186
RTECSNumber XU7525000
TSCA Yes