missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonylmethyl Chloride 98.0+%, TCI America™

Catalog No. T15055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15055G Supplier TCI America Supplier No. T15055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7569-26-8
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD00134281
InChI Key ZQPJNJKCPJDTIQ-UHFFFAOYSA-N
Synonym Chloromethyl p-Tolyl Sulfone, p-Tolylsulfonylmethyl Chloride
PubChem CID 562845
IUPAC Name 1-chloromethanesulfonyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CCl

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 5 g
Formula Weight 204.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonylmethyl Chloride