missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonylacetonitrile 99.0+%, TCI America™

Catalog No. T08465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08465G 5 g
T084625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08465G Supplier TCI America Supplier No. T08465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5697-44-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00039487
InChI Key BBNNLJMGPASZPD-UHFFFAOYSA-N
Synonym p-toluenesulfonylacetonitrile, 2-tosylacetonitrile, tosylacetonitrile, p-tolylsulfonylacetonitrile, 4-methylphenyl sulfonyl acetonitrile, 4-toluenesulfonylacetonitrile, 4-methylphenyl sulfonylacetonitrile, 4-toluenesulphonylacetonitrile, p-cyanomethylsulfonyl toluene, tosylmethyl isocyanide
PubChem CID 79776
IUPAC Name 2-(4-methylbenzenesulfonyl)acetonitrile
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 149°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 195.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonylacetonitrile