missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonic Acid Monohydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 106°C |
| Boiling Point | 140°C |
| Color | White |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| UN Number | 2585 |
| Formula Weight | 172.20 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | p-Toluenesulfonic Acid Monohydrate |
Safety and Handling
| EINECSNumber | (3)-1901 |
| RTECSNumber | DB7164000 |
| TSCA | Yes |