missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic Acid Monohydrate 98.0+%, TCI America™

Catalog No. T026725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T026725G 25 g
T0267500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T026725G Supplier TCI America Supplier No. T026725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00064387
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Melting Point 106°C
Boiling Point 140°C
Color White
Quantity 25 g
Physical Form Crystalline Powder
UN Number 2585
Formula Weight 172.20
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material p-Toluenesulfonic Acid Monohydrate
EINECSNumber (3)-1901
RTECSNumber DB7164000
TSCA Yes