missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic Acid Monohydrate, Reagents

Catalog No. 01337641
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337641 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337641 Supplier Labchem Inc Supplier No. C1341050500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

p-Toluenesulfonic Acid Monohydrate, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.21
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
IUPAC Name 4-methylbenzene-1-sulfonic acid hydrate
SMILES O.CC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Melting Point 106°C
Boiling Point 140°C
Color White
Quantity 500 g
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.24
Grade ACS
Chemical Name or Material p-Toluenesulfonic Acid Monohydrate