missing translation for 'onlineSavingsMsg'
Learn More

p-Toluamide 98.0+%, TCI America™

Catalog No. T125725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T125725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T125725G Supplier TCI America Supplier No. T125725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-55-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008002
InChI Key UHBGYFCCKRAEHA-UHFFFAOYSA-N
Synonym p-toluamide, benzamide, 4-methyl, p-methylbenzamide, 4-methyl-benzamide, 4-toluamide, 4-carbamoyltoluene, unii-n0v326v2f5, tolylsaureamid, n-toluamide, p-tolylamide
PubChem CID 69274
IUPAC Name 4-methylbenzamide
SMILES CC1=CC=C(C=C1)C(N)=O

Specifications

Melting Point 159°C
Color White
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluamide
EINECSNumber (3)-1407
RTECSNumber CV5570150
TSCA Yes